Specifications
- Size:500 Assays
- Cat. No.:103010-130
- Storage temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-71112
Specifications
About this item
Hydrogen peroxide is involved in a number of biological events, in particular, free radical-induced biochemical reactions.
SensoLyte® ADHP Hydrogen Peroxide Assay Kit uses highly purified ADHP to quantify hydrogen peroxide in solutions, in cell extracts and in live cells. It can also be used to detect a variety of oxidase activities through enzyme-coupled reactions. The kit includes ADHP phosphatase substrate (Ex/Em=573/590 nm upon oxidation) and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments, among other essential assay components.