- Size:500 Assays
- Cat. No.:103010-128
- Storage temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-71111
HRP conjugates are extensively used as secondary detection reagents in ELISAs, immuno-histochemical techniques and Northern, Southern and Western blot analyses.
SensoLyte® ADHP Peroxidase Assay Kit uses highly purified ADHP to quantify peroxidase activity in solutions, in cell extracts and in live cells and on solid surfaces (such as PVDF membranes). The kit can be used for characterizing kinetics of enzyme reaction and high throughput screening of oxidase inhibitors. The kit inlcludes ADHP peroxidase substrate (Ex/Em=573/590 nm upon oxidation) and an optimized ‘mix and read’ assay protocol that is compatible with HTS liquid handling instruments, among all essential assay components.