Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® NADP/NADPH Assay Kit (Colorimetric), AnaSpec
SensoLyte® NADP/NADPH Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-618
Supplier:  Anaspec
CAS Number:  
SensoLyte® NADP/NADPH Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-618
Supplier:  Anaspec
Supplier Number:  AS-72205
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-618
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72205

Specifications

About this item

Nicotinamide nucleotides are key players in the energy transforming and oxidation-reduction reactions of a cell

The SensoLyte® NADP/NADPH Assay Kit utilizes an optimized enzymatic recycling method for the quantification of NADP/NADPH without interfering with NAD/NADH. The final product of reaction, formazan, has blue color and can be detected at absorbance 565 nm. The intensity of the color produced is proportional to NADP/NADPH concentration.

NADP and NADPH act as allosteric effectors in modulating enzyme activities. The reducing power of NADPH is a required cofactor for enzymes that are pro-oxidants such as nitric oxide synthase and NADPH oxidase. NADP/NADPH ratio is involved in the control of the oxidative shunt. Each of these forms has a particular role during differentiation and similarity processes as well as being target functions in the cytotoxic action of ionizing radiations and alkylating agents.