Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Anti-MOG(35-55) IgG Quantitative ELISA Kit (Mouse/Rat), AnaSpec
SensoLyte® Anti-MOG(35-55) IgG Quantitative ELISA Kit (Mouse/Rat), AnaSpec
Catalog # 102997-814
Supplier:  Anaspec
CAS Number:  
SensoLyte® Anti-MOG(35-55) IgG Quantitative ELISA Kit (Mouse/Rat), AnaSpec
Catalog # 102997-814
Supplier:  Anaspec
Supplier Number:  AS-54465
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. No.:
    102997-814
  • Storage temperature:
    +4 °C
  • Includes:
    One 96-well plate, essential components and detailed protocol and references
  • Supplier no.:
    AS-54465

Specifications

About this item

MOG peptide fragment (35-55) induces autoantibody production and relapsing-remitting neurological disease causing extensive plaque-like demyelination

The SensoLyte® Anti-MOG (35-55) IgG Quantitative ELISA Kit (mouse/rat) provides a convenient, quantitative assay for detecting anti-MOG (35-55) IgG. This colorimetric assay, useful for determining the amount of anti-MOG (35-55) IgG present, can help provide information on the role anti-MOG (35-55) IgG plays in the development of EAE, an in vivo animal model for MS pathogenesis. The assays are performed in a convenient 96-well (strips format).

Autoantibody response to MOG (35-55) is present in multiple sclerosis (MS) patients and MOG (35-55)-induced experimental autoimmune encephalomyelitis (EAE) mice.