Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Green Glutaminyl Cyclase Activity Assay (Fluorimetric), AnaSpec Inc.
SensoLyte® Green Glutaminyl Cyclase Activity Assay (Fluorimetric), AnaSpec Inc.
Catalog # 103010-676
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® Green Glutaminyl Cyclase Activity Assay (Fluorimetric), AnaSpec Inc.
Catalog # 103010-676
Supplier:  Anaspec Inc
Supplier Number:  AS-72230
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-676
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72230

Specifications

About this item

The SensoLyte® Green Glutaminyl Cyclase Activity Assay Kit provides a convenient, two-step homogeneous procedure for measuring enzyme activity from various sources using a green fluorescence substrate

Many biologically active peptides and proteins have pyroglutamate (pGlu) as their first amino acid. This modification protects them from degradation by amino peptidases and in many cases determines functionality of protein or peptide. However, pyroglutamate modification sometimes may lead to negative side effects. For example, pGlu modified beta-amyloid peptides (pGlu-Aβ) aggregate much faster compared to non-pyroglutamic ones and form toxic oligomers that may damage neurons. Recent studies linked pGlu-Aβ to the Alzheimer’s Disease because it was identified as a major component of amyloid plaques. pGlu can be formed when glutamine or glutamic acid spontaneously convert into pyroglutamate. However, cyclic reaction rate is dramatically increased in the presence of Glutaminyl Cyclase (QC), also known as Glutaminyl-peptide Cyclotransferase (QPCT). Human Glutaminyl Cyclase has been isolated from nerve tissue, and detected in cerebrospinal fluid and saliva.

During the first step, substrate is incubated with Glutaminyl Cyclase or enzyme-containing samples and converted into the pyroglutamate form. Secondly, glutaminyl cyclase developer is added to remove pGlu residue and generate the green fluorophore. It can be detected with excitation at 490 nm and emission at 520 nm. Produced fluorescence is proportional to the enzyme activity. This kit is ideal for high throughput screening (HTS) of glutaminyl cyclase activators and inhibitors. The long wavelength of the green fluorophore is less interfered by autofluorescence of cell components and test compounds. Assay sensitivity is 1.5 ng/mL of active enzyme.