ContactUsLinkComponent
SensoLyte® Glutathione Cellular Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® Glutathione Cellular Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-520
 :  Anaspec
 :  
SensoLyte® Glutathione Cellular Assay Kit (Fluorimetric), AnaSpec Inc.
  103010-520
 :  Anaspec
 :  AS-72158
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    100 Assays
  • Cat. No.:
    103010-520
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72158
Glutathione (GSH) is an important antioxidant molecule that functions as a cofactor for a variety of enzymes and is involved in various biological processes
The SensoLyte® Glutathione Cellular Assay Kit utilizes an optimized enzymatic method for determination of the glutathione levels in cell or tissue extracts. The substrate provided in the kit can selectively react with GSH in the reaction catalyzed by glutathione S-transferase (GST) and form a fluorescent glutathione adduct. The resulting blue fluorescence can be monitored at excitation /emission= 390 nm/480 nm. The assay has a linear range of 0-100 µM of GSH. This kit does not require the usage of deproteination for biological samples.
A variation in glutathione levels is an indicator of oxidative stress.