Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Total GSH Assay Kit (Colorimetric), AnaSpec
SensoLyte® Total GSH Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-510
Supplier:  Anaspec
CAS Number:  
SensoLyte® Total GSH Assay Kit (Colorimetric), AnaSpec
Catalog # 103010-510
Supplier:  Anaspec
Supplier Number:  AS-72153
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. No.:
    103010-510
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72153

Specifications

About this item

Glutathione (GSH) is an important antioxidant molecule that functions as a cofactor for a variety of enzymes and is involved in various biological processes

The SensoLyte® Total GSH Quantitation Assay Kit utilizes the widely used Elmann’s reagent and an optimized enzymatic recycling method for the quantification of total glutathione. Glutathione reductase converts oxidized glutathione to reduced glutathione. Reduced GSH reacts with Ellman’s Reagent [5, 5’-dithiobis (2-nitrobenzoic acid)] and generates 2-nitro-5-thiobenzoic acid (TNB) producing a yellow color that can be detected at absorbance 405 to 415 nm. The intensity of the color produced is proportional to GSH concentration. The kit also provides 5-sulfosalicylic acid for protein removal from biological samples.

Total GSH includes reduced GSH and oxidized GSH (GSSG). The concentrations of GSH and GSSG and their molar ratio can be used for indicating the oxidative stress status.