Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-626
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 Enterokinase Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-626
Supplier:  Anaspec
Supplier Number:  AS-72209
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-626
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72209

Specifications

About this item

Enterokinase is a heterodimeric serine protease produced by cells in the duodenal wall

The SensoLyte® 520 Enterokinase Assay Kit is optimized for detecting activity of enterokinase. This kit employs a novel internally quenched 5-FAM/QXL FRET substrate for enterokinase. Enterokinase cleaves the FRET substrate into two separate fragments resulting in release of 5-FAM fluorescence which can be monitored at excitation/emission= 490/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples and test compounds. The assay can detect as low as 1.25 ng/mL of active enterokinase.

It converts trypsinogen to trypsin, indirectly activating a number of pancreatic digestive enzymes. Enterokinase is a key enzyme for intestinal digestion of proteins. Its deficiency may cause severe protein malabsorption. Enterokinase also plays an important role in acute experimental and clinical pancreatitis. The high degree of specificity exhibited by enterokinase for cleaving affinity or other tags from recombinant proteins makes it an ideal tool for use in biochemical applications.