Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Green Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® Green Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-566
Supplier:  Anaspec
CAS Number:  
SensoLyte® Green Elastase Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-566
Supplier:  Anaspec
Supplier Number:  AS-72178
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. No.:
    103010-566
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72178

Specifications

About this item

The SensoLyte® Green Elastase Assay Kit is a FRET-based assay that detects elastase activity. The kit provides an elastin, natural substrate for elastases, labeled with 5-FAM fluorophore and QXL®520 quencher. Elastase-catalyzed hydrolysis yields brightly green fluorescence. Increase in fluorescence intensity is directly proportional to enzyme activity. The kit does not require any separation steps and can be used to continuously measure the kinetics of elastase.