Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 ECEs Activity Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 ECEs Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103008-980
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® 520 ECEs Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103008-980
Supplier:  Anaspec Inc
Supplier Number:  AS-72243
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103008-980
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72243

Specifications

About this item

ECEs or Endothelin converting enzymes (ECE-1/ECE-2) are known as rate-limiting enzymes for the formation of the vasoconstrictor peptide endothelin-1 (ET-1) from its precursor proendothelin-1

The SensoLyte® 520 ECEs Activity Assay Kit can be used to detect ECE activity in biological samples or in purified enzyme preparations. The substrate provided in this kit was based on a beta amyloid fragment. When active ECEs cleave the FRET substrate, it results in an increase of 5-FAM fluorescence, and is monitored at excitation/emission = 490/520 nm. The long wavelength fluorescence of 5-FAM shows less interference from autofluorescence of components in biological samples and test compounds. This assay can detect as low as 15.6 ng/mL active ECE-1 and 62.5 ng/mL active ECE-2.

ECE-1 can also degrade amyloid ß fragments both in vitro and in vivo. Highly homologous and structurally comparable to ECE-1, ECE-2 was shown in homozygous knockout mice to have decreased Aß degradation and significant elevated Aß 1-40 and Aß 1-42 in the brain.