Specifications
- Size:100 Assays
- Cat. No.:103010-600
- Storage temperature:– 78 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-72196
Specifications
About this item
DPPIV is a widely distributed serine protease that cleaves two amino acids from small peptides containing alanine or proline in the second position of the N-terminus of the peptide
The SensoLyte® Rh110 DPPIV Assay Kit provides a convenient method for high throughput screening of DPPIV inhibitors and for continuous assay of DPPIV activity utilizing a fluorogenic peptide. Upon enzyme cleavage, the peptide releases the Rh110 fluorophore with bright green fluorescence that can be detected at Ex/Em=490/520 nm. The longer-wavelength spectra and higher extinction coefficient of the Rh110 provide greater sensitivity and less interference from reaction components.
DPPIV activity is especially critical for inactivation of GLP-1, a potent antihyperglycemic hormone, inducing glucose-dependent stimulation of insulin secretion. DPPIV inhibitors provide a potent treatment for type 2 diabetes prolonging the activity of GLP-1 and improving insulin secretion and blood glucose control regulation.