Specifications
- Size:500 Assays (96-well format) or 1250 Assays (384-well format)
- Cat. No.:103010-138
- Storage temperature:– 20 °C
- Includes:The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-71118
Specifications
About this item
Central to the execution phase of apoptosis are the two closely related caspase-3 and caspase-7
The SensoLyte® Homogeneous AMC Capase-3/7 Assay Kit uses Ac-DEVD-AMC as the fluorogenic indicator for assaying caspase-3/7 activities. Upon caspase-3/7 cleavage, Ac-DEVD-AMC generates the AMC fluorophore which has bright blue fluorescence and can be detected at Ex/Em=354 nm/442 nm. A bi-function assay buffer in this kit is designed to lyze the cells and measure the enzyme activity at the same time. Thus, this kit can measure caspase-3/7 activity in cell culture directly in a 96-well or 384-well plate without a time-consuming cell extraction step. In case the cells are cultured in larger plates or flasks, a lysis buffer and protocol for cell lysate preparation are also conveniently included in the kit. The kit is suitable for high throughput screening of apoptosis inducers and inhibitors.
They share common substrate specificity and structure, but differ completely in the sequence of their respective N-terminal regions. Both caspase-3 and caspase-7 have substrate selectivity for the amino acid sequence Asp-Glu-Val-Asp (DEVD).