- Size:100 Assays
- Cat. No.:103010-506
- Storage temperature:– 78 °C
- Includes:The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
- Supplier no.:AS-72150
The calpains are a family of intracellular Ca2+ supdependent cysteine proteases
The SensoLyte® AMC Calpain Assay Kit is optimized for detecting calpain activities. This kit contains a fluorogenic peptide substrate, suc-LLVY-AMC. Upon calpain protease cleavage, suc-LLVY-AMC generates the AMC fluorophore emitting bright blue fluorescence and can be monitored at excitation/emission=354/442 nm. Increase in AMC fluorescence is proportional to calpain activity. The assay can detect both calpain 1 (μm) and 2 (m) activities and is ideal for kinetic study of these enzymes. The assays are performed in a convenient 96-well microplate format.
The best-characterized calpains are isoforms 1 (µm) and 2 (m). Calpains respond to Ca2+ signals by cleaving many specific proteins, thereby irreversibly modifying their function(s). They are implicated in a variety of Ca2+ regulated cellular processes as well as various pathological phenomena, such as ischemic injury, muscular dystrophy, diabetes, cataract, atherosclerosis, Alzheimer's disease, and cancer. Calpains represent potential therapeutic targets for drug discovery.