Specifications
- Size:96 Assays
- Cat. No.:103001-640
- Storage temperature:+4 °C
- Includes:Kit includes a 96-well plate, essential components and detailed protocol and references
- Supplier no.:AS-55554
Specifications
About this item
This SensoLyte® high-sensitivity (2pg/mL) beta-Amyloid (1-42) Quantitative ELISA Kit (Mouse/Rat) provides a convenient and quantitative assay for determining mouse/rat beta-Amyloid (1-42) (Aβ42) amount in cell and tissue lysate as well as in body fluids
Alzheimer's Disease (AD) is the most common neurodegenerative disorder in elderly people. It has been demonstrated that AD has biological causes and is characterized by the presence of senile plaques and neurofibrillary tangles mainly in cerebral cortex and hippocampus brain regions. Beta-Amyloid (1-40) (Aβ40) and beta-Amyloid (1-42) (Aβ42) are the main components of the above plaques; however, other forms of beta-Amyloid peptides are also present. Both peptides are cleaved from the Amyloid Precursor Protein (APP) by β-secretase and γ-secretase enzymes. Many studies suggest that Aβ42 or/and Aβ43 are required to initiate formation of amyloid plaques and neurofibrills that leads to the neurodegeneration,while Aβ40 is less neurotoxic.
Compared to other mouse/rat anti-Aβ42 ELISA kits on the market, it takes less time to run this assay. HRP conjugated detection antibody in this kit is added simultaneously with the samples and standards during the assay. This eliminates extra incubation and washing steps and makes this kit a one-step procedure for Aβ42 quantification.