Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® Anti-Mouse/ Rat β-Amyloid (1-40) Quantitative ELISA (Colorimetric), AnaSpec
SensoLyte® Anti-Mouse/ Rat β-Amyloid (1-40) Quantitative ELISA (Colorimetric), AnaSpec
Catalog # 103001-638
Supplier:  Anaspec
CAS Number:  
SensoLyte® Anti-Mouse/ Rat β-Amyloid (1-40) Quantitative ELISA (Colorimetric), AnaSpec
Catalog # 103001-638
Supplier:  Anaspec
Supplier Number:  AS-55553
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. No.:
    103001-638
  • Storage temperature:
    +4 °C
  • Includes:
    Kit includes a 96-well plate, essential components and detailed protocol and references
  • Supplier no.:
    AS-55553

Specifications

About this item

This SensoLyte® high-sensitivity (2pg/mL) beta-Amyloid (1-40) Quantitative ELISA Kit (Mouse/Rat) provides a convenient and quantitative assay for determining mouse/rat beta-Amyloid (1-40) (Aβ40) amount in cell and tissue lysate as well as in body fluids

Alzheimer's Disease (AD) is the most common neurodegenerative disorder in elderly people. It has been demonstrated that AD has biological causes and is characterized by the presence of senile plaques and neurofibrillary tangles mainly in cerebral cortex and hippocampus brain regions. Beta-Amyloid (1-40) (Aβ40) and beta-Amyloid (1-42) (Aβ42) are the main components of the above plaques; however, other forms of beta-Amyloid peptides are also present. Both peptides are cleaved from the Amyloid Precursor Protein (APP) by β-secretase and γ-secretase enzymes. Many studies suggest that Aβ42 or/and Aβ43 are required to initiate formation of amyloid plaques and neurofibrills that leads to the neurodegeneration, while Aβ40 is less neurotoxic.

Compared to other mouse/rat anti-Aβ40 ELISA kits on the market, it takes less time to run this assay. HRP conjugated detection antibody in this kit is added simultaneously with the samples and standards during the assay. This eliminates extra incubation and washing steps and makes this kit a one-step procedure for Aβ40 quantification.