Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 ADAMTS13 Activity Assay (Fluorimetric), AnaSpec
SensoLyte® 520 ADAMTS13 Activity Assay (Fluorimetric), AnaSpec
Catalog # 103010-680
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 ADAMTS13 Activity Assay (Fluorimetric), AnaSpec
Catalog # 103010-680
Supplier:  Anaspec
Supplier Number:  AS-72232
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    96 Assays
  • Cat. No.:
    103010-680
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72232

Specifications

About this item

ADAMTS13 (A Disintegrin-like and Metalloprotease with Thrombospondin type-1 motifs 13) is a metalloproteinase that is primarily synthesized in liver cells and secreted into blood circulation

The SensoLyte® 520 ADAMTS13 Assay Kit employs an internally quenched vWF73 FRET peptide substrate for the detection of enzyme activity. This substrate contains a novel 5-FAM/QXL® 520 FRET pair. ADAMTS13 cleaves this FRET substrate into two separate fragments resulting in the release of 5-FAM fluorescence, which can be monitored at Ex/Em= 490/520 nm. The long wavelength fluorescence of 5-FAM is less interfered by the autofluorescence of components in biological samples.

ADAMTS13 specifically cleaves von Willebrand factor (vWF) at the Tyr1605-Met1606 peptide bond. vWF is a multimeric glycoprotein that mediates adhesion and aggregation of platelets. ADAMTS13 is essential for preventing platelet aggregation in the circulation. Deficiency of plasma ADAMTS13 activity may lead to thrombotic thrombocytopenic purpur (TTP), a potentially lethal syndrome characterized by the formation of VWF-platelet-rich thrombi in the arterioles and capillaries. Studies have shown that ADAMTs13 is also involved in angiogenesis, inflammation, atherosclerosis, and liver cirrhosis. Measurement of ADAMTS13 activity in plasma has been used for supporting diagnosis of acute TTP.