Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 ADAM10 Activity Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 ADAM10 Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-668
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 ADAM10 Activity Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-668
Supplier:  Anaspec
Supplier Number:  AS-72226
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-668
  • Storage temperature:
    – 78 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72226

Specifications

About this item

ADAM10 has been shown to be an effective α-secretase in vitro and in vivo

The SensoLyte® 520 ADAM10 Activity Assay Kit is an optimized assay that can be used to detect the ADAM 10 activity. The unique FRET substrate contained in the kit was designed to reduce the cross reactivity with ADAM17 (also called tumor necrosis factor- α-converting enzyme, TACE). When active ADAM10 cleaves the FRET substrate, it results in an increase of 5-FAM fluorescence monitored at excitation/emission = 490/520 nm. The long wavelength fluorescence of 5-FAM is also less interfered by the autofluorescence of components in biological samples and test compounds. This assay can detect as low as 0.25 ng/mL active ADAM10.

This enzyme acts as a sheddase to cleave cell surface proteins involved in neuropathology, inflammatory response and tumor progression.