Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 390 ACE2 Activity Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® 390 ACE2 Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103008-964
Supplier:  Anaspec Inc
CAS Number:  
SensoLyte® 390 ACE2 Activity Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103008-964
Supplier:  Anaspec Inc
Supplier Number:  AS-72086
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103008-964
  • Storage temperature:
    – 20 °C
  • Includes:
    The assays can be performed in a convenient 96-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-72086

Specifications

About this item

ACE2 or ACEH, angiotensin-converting enzyme 2, a homolog of angiotensin-converting enzyme, is a novel zinc metallopeptidase with specificity, tissue distribution, and function distinct from those of ACE

The SensoLyte® 390 ACE2 Activity Assay Kit provides a convenient assay for high throughput screening of ACE2 enzyme inhibitors and inducers using a Mc-Ala/Dnp fluorescence resonance energy transfer (FRET) peptide. In the FRET peptide, the fluorescence of Mc-Ala is quenched by Dnp. Upon cleavage into two separate fragments by ACE2, the fluorescence of Mc-Ala is recovered, and can be monitored at excitation/emission = 330/390 nm. This assay can detect the activity of subnanogram level of ACE2. Assays are performed in a convenient 96-well microplate format.

ACE2 may play a unique role in the renin-angiotensin system and mediate cardiovascular and renal function.