Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 MMP-13 Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 MMP-13 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-242
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 MMP-13 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-242
Supplier:  Anaspec
Supplier Number:  AS-71156
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-242
  • Storage temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71156

Specifications

About this item

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components

The SensoLyte® 520 MMP-13 Assay Kit uses a 5-FAM (fluorophore) and QXL® 520 (quencher) labeled FRET peptide substrates for continuous measurement of the enzyme activities. In an intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL® 520. Upon the cleavage of the FRET peptide by MMP-13, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL® 520 based FRET peptide is less interfered by the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The assays are performed in a convenient 96-well or 384-well microplate format. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-13 activity, please choose the SensoLyte® Plus MMP-13 Assay Kit.

MMP-13 (collagenase-3, rat collagenase) is involved in quite a few diseases such as arthritis and asthma.