Order Entry
Puerto Rico
ContactUsLinkComponent
 
SensoLyte® 520 MMP-7 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-236
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 MMP-7 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-236
Supplier:  Anaspec
Supplier Number:  AS-71153
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays (96-well) or 250 Assays (384-well)
  • Cat. No.:
    103010-236
  • Storage temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71153

Specifications

About this item

MMP-7 (matrilysin, PUMP) is involved in biological events, such as tissue remodeling, cell proliferation and host defenses.

SensoLyte® 520 MMP-7 Assay Kit uses a 5-FAM (fluorophore) and QXL520™ (quencher) labeled FRET peptide substrates for continuous measurement of the enzyme activities. In an intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL520™. Upon the cleavage of the FRET peptide by MMP-7, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL520™ based FRET peptide is less interfered by the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The assays are performed in a convenient 96-well or 384-well microplate format.