Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 520 MMP-2 Assay Kit (Fluorimetric), AnaSpec
SensoLyte® 520 MMP-2 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-232
Supplier:  Anaspec
CAS Number:  
SensoLyte® 520 MMP-2 Assay Kit (Fluorimetric), AnaSpec
Catalog # 103010-232
Supplier:  Anaspec
Supplier Number:  AS-71151
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    100 Assays
  • Cat. No.:
    103010-232
  • Storage temperature:
    –20 °C
  • Includes:
    The assays can be performed in a convenient 96-well or 384-well microplate format, kit includes essential components and detailed protocol and references
  • Supplier no.:
    AS-71151

Specifications

About this item

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated proteins capable of digesting extracellular matrix components

SensoLyte® 520 MMP-2 Assay Kit uses a 5-FAM (fluorophore) and QXL520™ (quencher) labeled FRET peptide substrate for continuous measurement of the enzyme activities. In an intact FRET peptide, the fluorescence of 5-FAM is quenched by QXL520™. Upon the cleavage of the FRET peptide by MMP-2, the fluorescence of 5-FAM is recovered, and can be continuously monitored at excitation/emission = 490 nm/520 nm. With superior fluorescence quantum yield and longer emission wavelength, 5-FAM/QXL520™ based FRET peptide is less interfered by the autofluorescence of test compounds and cellular components and provides better assay sensitivity. The assays are performed in a convenient 96-well or 384-well microplate format.

The importance of MMPs in tumor development and invasion is well known. MMP-2 (gelatinase A) is proposed as a potential therapeutic target.