Order Entry
Puerto Rico
ContactUsLinkComponent
SensoLyte® 490 MMP-1 Assay Kit (Fluorimetric), AnaSpec Inc.
SensoLyte® 490 MMP-1 Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-150
Supplier:  Anaspec
CAS Number:  
SensoLyte® 490 MMP-1 Assay Kit (Fluorimetric), AnaSpec Inc.
Catalog # 103010-150
Supplier:  Anaspec
Supplier Number:  AS-71128
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    500 Assays
  • Cat. No.:
    103010-150
  • Storage temperature:
    –20 °C
  • Includes:
    Kit includes a 96-well plate, essential components and detailed protocol and references
  • Supplier no.:
    AS-71128

Specifications

About this item

MMP-1 (interstitial collagenase, fibroblast collagenase) is an extracellular protease

The SensoLyte® 490 MMP-1 Assay Kit uses an optimized fluorogenic peptide substrate for continuous measurement of the enzyme activity, providing speed, convenience and accuracy in analyzing MMP-1 and screening MMP-1 inhibitors. Members of the MMP family have poor substrate sequence specificity, making it difficult to use a peptide substrate alone to differentiate the activity of a particular MMP from other MMPs. If several MMPs are coexisting in your samples and you would like to specifically measure MMP-1 activity, please choose the SensoLyte® Plus MMP-1 Assay Kit.

This enzyme plays a significant role in some diseases and has been identified as a drug screening target.