Frits for Omnifit® EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns, Diba Industries
Supplier: Antyila Scientific
Certificates
Frits for Omnifit® EZ, EZ SolventPlus™, BenchMark™, HiT™, and SolventPlus™ Columns, Diba Industries
Supplier: Antyila Scientific
About this item
Select from a range of porosities for your media and chemistry.
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Sodium hydroxide 0.5 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Screw Top Vials and Caps, 2 ml, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
VWR® Pipette Controller
Supplier: VWR International
Closures for 2 ml Screw Top Vials, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Coned Port Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Gas Clean Purification System, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Instrument Supplies for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Captiva Syringe Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø = ¹/₁₆" (1.6 mm), IDEX Health & Science
Supplier: Upchurch Scientific
J.T.Baker®, Phase Separation Paper
Supplier: AVANTOR PERFORMANCE MATERIALS US
Stainless Steel Frits, Upchurch Scientific®
Supplier: Upchurch Scientific