Order Entry
United States
ContactUsLinkComponent
Heater Element, Merit Water Still, Dynalon
Heater Element, Merit Water Still, Dynalon
Catalog # 490012-278
Supplier:  Dynalon
undefined
Heater Element, Merit Water Still, Dynalon
Catalog # 490012-278
Supplier:  Dynalon
Supplier Number:  540014-0001

Specifications

  • Description:
    Heater element for QWS4 Water Still
  • Environmentally Preferable:
  • Cat. No.:
    490012-278
  • Electrical:
    230 V
  • Supplier no.:
    540014-0001

Specifications

About this item

Replacement Heater Element for economic Merit Water Still.

The Merit Water Still combines economy and high performance with a host of additional features that comparable stills cannot match. The Merit Water Still is the ideal choice for the budget-conscious laboratory that cannot afford to compromise quality.

Dimension: 12.7L×12.7W×36.83H cm.