Wavelength and UV Photometric Accuracy Standard, FireFlySci
Catalog # 76016-704
Supplier: FIREFLYSCI, INC.
CAS Number:
Specifications
- Description:Wavelength and UV Photometric Accuracy Standard
- Cat. No.:76016-704
Specifications
About this item
Wavelength accuracy is determined by scanning a wavelength filter with sharp, clearly defined transmission peaks and comparing the results to those on the certificate of calibration provided
- Scratch-resistant coating protects optical surfaces
- Patented nano-deposition manufacturing process means that filters never have to be recalibrated if used properly
- Wavelength accuracy in the UV and VIS range (329 – 875 nm)
- Photometric accuracy in the UV range (270 – 340 nm)
Its highly accurate and consistent spectrophotometer calibration every time. It can be used in any spectrophotometer (with standard 12.5 x 12.5 x 45 mm holder).
Certifications: Complies to NIST.
Delivery: Supplied with instructions, certificate of calibration with NIST traceability and photometric and wavelength data tables.