Secondary Medication Sets, Baxter
About this item
Secondary medication set with male Luer lock adapter.
- Overall length: 94 cm (37")
- Drops capacity: 10/mL
: Supplied with hanger for primary solution container.
3 Options Available Below
Product Details & Documents
Secondary medication set with male Luer lock adapter.
- Overall length: 94 cm (37")
- Drops capacity: 10/mL
: Supplied with hanger for primary solution container.
Certificates
Search by Lot NumberProduct Family Options
Product Information
- AvailabilityPrice
- 68000-926Secondary medication set with interlink lever lock cannula (detached)Secondary medication set with interlink lever lock cannula (detached)Log in for availabilityCase of 48$414.97Supplier Number: 2C7451
- 95037-754Secondary medication setSupplier Number: 2C7461
- 47732-816Secondary medication set with Duo-vent spike and interlink lever lock cannula (detached)Secondary medication set with Duo-vent spike and interlink lever lock cannula (detached)Log in for availabilityCase of 48$497.78Supplier Number: 2C7452