These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
These tweezers have an anti-magnetic WA1 handle with replaceble 159-BUIS tips made of boxwood (Buxus). Tips are attached with screws. Torque: cN.m 9 ±1.
Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox. Length is 130mm.