About this item
These precision tweezers from Dumont made of high quality steels with perfect finish suits all application needs.
The anti-magnetic steel ergonomic tweezers have a soft electro static discharge (ESD) grip. The red rubber handle has very high resistivity and a comfortable grip. Antimagnetic (80%) contains C, Cr, Ni and Mo and has good resistance to corrosion due to a high concentration of molybdenum but is not as hard as Inox. Length is 115 mm.