Order Entry
Puerto Rico
ContactUsLinkComponent
Numbered Gunshot Residue Sampler, Electron Microscopy Sciences
Numbered Gunshot Residue Sampler, Electron Microscopy Sciences
Catalog # 103306-270
undefined
Numbered Gunshot Residue Sampler, Electron Microscopy Sciences
Catalog # 103306-270
Supplier Number:  76480-16
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Numbered Gunshot Residue Sampler
  • Cat. No.:
    103306-270
  • Supplier no.:
    76480-16

Specifications

About this item

SEM mounts with adhesive tab secures in glass vial to avoid contamination.

  • Certified aluminum SEM mount
  • Specify 100 consecutive numbers between 0 and 9999
  • Vials are 25 mm OD × 55 mm high

Numbered, certified aluminum mount with a double-coated adhesive carbon tab applied to secure the sample in a clean, glass vial 25 mm OD x 55 mm high (1 × 2.17"). Packaged in a box of 100. Numbers can be specified as 100 consecutive numbers between 0 and 9999 (greatest numbers). Please inform customer service about the numbering sequence when ordering this product.