Order Entry
Puerto Rico
ContactUsLinkComponent
AnaTag™ APC Labeling Kit, AnaSpec
AnaTag™ APC Labeling Kit, AnaSpec
Catalog # 103010-444
Supplier:  Anaspec
CAS Number:  
AnaTag™ APC Labeling Kit, AnaSpec
Catalog # 103010-444
Supplier:  Anaspec
Supplier Number:  AS-72111
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 Labeling Reaction
  • Cat. No.:
    103010-444
  • Storage temperature:
    +4 °C
  • Includes:
    This kit is optimized for conjugating allophycocyanin (APC) to antibody providing ample materials to perform one conjugation reaction for labeling up to 1 mg antibody
  • Supplier no.:
    AS-72111

Specifications

About this item

The AnaTag™ APC Labeling Kit is optimized for use in the conjugation of allophycocyanin (APC) to antibodies. APC is an ultra-sensitive, near-infrared fluorescent tracer with a high quantum yield (Ex/Em = 650 nm/660 nm). APC- labeled antibodies are used in applications such as flow cytometry and immunofluorescence. The AnaTagTM APC Labeling Kit contains SH-reactive APC. SMCC modified allophycocyanin reacts with the thiol groups of target antibody without the need for additional activation, thus simplifying the conjugation protocol. AnaTagTM APC Labeling Kit contains a chemically cross-linked APC (CL-APC) that is much more stable than the native APC, but still retains the original spectroscopic properties. The amount of APC supplied in this kit is sufficient for labeling up to 1 mg of antibody. The kit provides all reagents, purification columns and a detailed step-by-step protocol. Cross-linked and SMCC-activated APC are also available as separate products.