Order Entry
Puerto Rico
ContactUsLinkComponent
AnaTag™ HiLyte™ Fluor 647 Protein Labeling Kit, Ultra Convenient [3 labeling reactions], AnaSpec
AnaTag™ HiLyte™ Fluor 647 Protein Labeling Kit, Ultra Convenient [3 labeling reactions], AnaSpec
Catalog # 103010-358
Supplier:  Anaspec
CAS Number:  
AnaTag™ HiLyte™ Fluor 647 Protein Labeling Kit, Ultra Convenient [3 labeling reactions], AnaSpec
Catalog # 103010-358
Supplier:  Anaspec
Supplier Number:  AS-72049
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    3 Labeling Reactions
  • Cat. No.:
    103010-358
  • Storage temperature:
    +4 °C
  • Includes:
    The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG
  • Supplier no.:
    AS-72049

Specifications

About this item

HiLyte™ Fluor 647 acid, SE (Ex/Em=647 nm/679 nm) is an excellent fluorescent labeling dye that generates the protein conjugates that are slightly red-shifted compared to those of Cy® 5 dyes, resulting in an optimal match to filters designed for Cy®5 dyes

The HiLyte™ Fluor 647-protein conjugates are suitable for immunofluorescent staining, in situ hybridization, flow cytometry and other biological applications. Additionally, our in-house data indicated that HiLyte™ Fluor 647 is superior to Cy™5 in fluorescence polarization-based assays. The kit has all essential components for performing the conjugation reaction and for purifying the HiLyte™ Fluor 647-protein conjugate. The entire process takes only half an hour. The kit is able to perform three conjugation reactions with each reaction labeling up to 5 mg IgG. Kit Size: 3 labeling reactions (3 x 5 mg protein).

However, total fluorescence of the secondary antibody conjugates of HiLyte™ Fluor 647 dyes is significantly higher than that of Cy®5 conjugates supplied by other companies. Also, unlike Cy®5 dyes, HiLyte™ Fluor 647 dyes have very little change in absorption or fluorescence spectra when conjugated to most proteins, oligonucleotides and nucleic acids, thus yielding greater total fluorescence at the same degree of substitution.