To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Control
- Clonality:Monoclonal
- Clone:LTF-2
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2b
- Reactivity:Mouse
- Size:100 μg
- Environmentally Preferable:
- Form:liquid
- Cat. No.:75842-460
- Supplier no.:41823-77-100
Specifications
About this item
The LTF-2 monoclonal antibody is used as an Isotype Control immunoglobulin for rat IgG2b antibodies.
Type: Control
Antigen:
Clonality: Monoclonal
Clone: LTF-2
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse