To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Monocyte chemotactic protein 1
- Antigen Symbol:MCP-1
- Clonality:Monoclonal
- Clone:2H5
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Human
- Size:500ug
- Environmentally Preferable:
- Format:SAFIRE Purified
- Form:liquid
- Antigen Synonyms:CCL-2|MCAF|SCYA2|Monocyte chemotactic protein 1
- Cat. No.:75843-446
- Supplier no.:80111-25-500
The 2H5 monoclonal antibody specifically reacts with mouse, rat, human MCP-1 (Monocyte Chemoattractant Protein-1), also known as CCL2 and MCAF (Monocyte Chemotactic and Activating Factor). MCP-1 is a C-C (beta) chemokine that regulates cytokine production in monocytes and the expression of adhesion molecules. The 2h5 antibody can neutralize the bioactivity of recombinant or natural MCP-1.
Type: Primary
Antigen: MCP-1
Clonality: Monoclonal
Clone: 2H5
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Human