Order Entry
United States
ContactUsLinkComponent
Anti-Ly-6G Rat Monoclonal Antibody (BG Violet 450) [clone: RB6-8C5]
Anti-Ly-6G Rat Monoclonal Antibody (BG Violet 450) [clone: RB6-8C5]
Catalog # 75842-552
Anti-Ly-6G Rat Monoclonal Antibody (BG Violet 450) [clone: RB6-8C5]
Catalog # 75842-552
Supplier Number:  83122-40-100
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    Ly-6G
  • Clonality:
    Monoclonal
  • Clone:
    RB6-8C5
  • Conjugation:
    BG Violet 450
  • Flow Cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Size:
    100 μg
  • Environmentally Preferable:
  • Form:
    liquid
  • Antigen Synonyms:
    Ly6G|Gr1
  • Cat. No.:
    75842-552
  • Supplier no.:
    83122-40-100

Specifications

About this item

The RB6-8C5 monoclonal antibody reacts with the mouse Ly-6G (also known as Gr-1). The Ly-6G protein is a myeloid differentiation antigen of 21-25 kDa, expressed in a regulated manner by the myeloid lineage in the bone marrow, where the level of antigen expression is correlated with the granulocyte maturation and differentiation. In the bone marrow, the antigen is not expressed by the erythroid cells. From the peripheric cells, RB6-8C5 binds with monocytes, neutrophils and eosinophils.As a marker for the mouse monocytes, macrophages and granulocytes, the RB6-8C5 antibody is usually combined with M1/70, a macrophage labeling antibody (Anti-CD11b), for phenotypic analysis.

Type: Primary
Antigen: Ly-6G
Clonality: Monoclonal
Clone: RB6-8C5
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse