To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Interferon gamma
- Antigen Symbol:IFNG
- Clonality:Monoclonal
- Clone:XMG1.2
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:500 μg
- Environmentally Preferable:
- Format:SAFIRE Purified
- Form:liquid
- Antigen Synonyms:Type II interferon|T cell interferon|Interferon-g|Macrophage-activating factor (MAF)|IFNg|IFN-g|Interferon-γ|Immune interferon
- Cat. No.:75843-468
- Supplier no.:80812-25-500
Specifications
About this item
The XMG1.2 is a neutralizing antibody that binds with the mouse Interferon-gamma (IFN-γ) protein, a 15 -17 kDa cytokine with significant antibacterial, antiviral, and antitumoral properties. When secreted by natural killer cells and by natural killer T lymphocytes, it regulates the immune response and supports adaptive immunity when produced by Th1 or CD8+ T lymphocytes. IFN-γ plays an important role in the activation, the growth, and the differentiation of the macrophages, B and T lymphocytes, and natural killer cells. It interacts synergically with other cytokines, such as TNF-α, to inhibit proliferation of normal and transformed cells. IFN- γ is the primary cytokine that defines Th-1 cells.The biological activity of IFN-γ is not affected by glycosylation.
Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: XMG1.2
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse