To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Cytotoxic T-Lymphocyte Antigen 4
- Antigen Symbol:CTLA4
- Clonality:Monoclonal
- Clone:BNI3
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:500 μg
- Environmentally Preferable:
- Format:SAFIRE Purified
- Form:liquid
- Antigen Synonyms:Cytotoxic T-Lymphocyte Antigen 4|CTLA4|BNI3|CD152
- Cat. No.:75844-394
- Supplier no.:14611-25-500
Specifications
About this item
The BNI3 monoclonal antibody specifically reacts with human CD152, the Cytotoxic T-Lymphocyte Antigen 4 (CTLA-4). CTLA-4 is expressed on activated CD28+ T cells, and binds the B7 family members B7-1 (CD80) and B7-2 (CD86). The structure of CTLA-4 is similar to the structure of CD28, but the two molecules seem to have opposite roles on the T lymphocytes. CTLA-4 inhibits the progression of T cell activation, while CD28 stimulates it. This result explains the stimulating role that the immobilization of BNI3 plays on the T lymphocytes proliferation induced by CD28.
Type: Primary
Antigen: CTLA4
Clonality: Monoclonal
Clone: BNI3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human