Specifications
- Antibody Type:Primary
- Antigen Name:Cluster of Differentiation 8a
- Antigen Symbol:CD8a
- Clonality:Monoclonal
- Clone:RPA-T8
- Conjugation:BG Violet 450
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:T8|CD8 alpha|Ly-2|Lyt-2|Ly-35|Ly-B
- Cat. No.:75843-062
- Supplier no.:10131-40-25
Specifications
About this item
The RPA-T8 monoclonal antibody specifically reacts with the human CD8a molecule, a 32 kDa cell surface receptor expressed either as a heterodimer (CD8 α/β) or as a homodimer (CD8 α/α) on the majority of thymocytes, a subpopulation of mature T cells, and natural killer cells. CD8 interacts with the major histocompatibility complex class I (MHC class I) molecules on antigen-presenting cells or epithelial cells. The RPA-T8 antibody reacts with 13-48% of peripheral lymphocytes, 80% of thymocytes, and a subset of natural killer cells. RPA-T8, OKT8, and HIT8a antibodies do not compete with each other for binding to peripheral leukocytes, meaning that that they do not recognize the same epitope or block each other by steric hindrance.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD8a
Clonality: Monoclonal
Clone: RPA-T8
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human