Anti-CD69 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: FN50]
Catalog # 75843-094
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Name:Cluster of Differentiation 69
- Antigen Symbol:CD69
- Clonality:Monoclonal
- Clone:FN50
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen Synonyms:EA1|MLR-3|AIM|GP32/28|CD69 molecule|CLEC2C|BL-AC/P26
- Cat. No.:75843-094
- Supplier no.:11711-60-100
Specifications
About this item
The FN50 monoclonal antibody specifically reacts with human CD69, the 27-33 kDA type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: FN50
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human