Anti-CD69 Armenian Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: H1.2F3]
75843-104
- Antibody Type:Primary
- Antigen Name:Cluster of Differentiation 69
- Antigen Symbol:CD69
- Clonality:Monoclonal
- Clone:H1.2F3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Antigen Synonyms:EA1|MLR-3|AIM|GP32/28|CD69 molecule|CLEC2C|BL-AC/P26
- Cat. No.:75843-104
- Supplier no.:11712-50-100
The H1.2F3 monoclonal antibody specifically reacts with human CD69, the 27-33 kDA type II transmembrane protein also known as the very early activation antigen (VEA) or the activation inducer molecule (AIM). It is expressed as a disulfide-linked dimer on B cells, T cells, NK cells, platelets, eosinophils, and neutrophils. It increases in expression upon cell activation and seems to serve a role as a signaling receptor.
Type: Primary
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse