Anti-B3GAT1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: TBO1]
Catalog # 75842-028
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Name:Beta-1,3-glucuronyltransferase 1
- Antigen Symbol:B3GAT1
- Clonality:Monoclonal
- Clone:TBO1
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgM
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:LEU7|GLCATP|CD57|GLCUATP|NK-1|NK|HNK1
- Cat. No.:75842-028
- Supplier no.:09611-50-25
Specifications
About this item
The TBO1 monoclonal antibody specifically binds to human CD57, a 110 kDA glycoprotein expressed on a subset of natural killer lymphocytes, cells, neural cells and striated muscle. It is reported that the molecule is involved in cell-matrix interactions and is upregulated in some disease states.
Type: Primary
Antigen: B3GAT1
Clonality: Monoclonal
Clone: TBO1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human