Anti-CD5 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: UCHT2]
Catalog # 75843-040
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD5
- Clonality:Monoclonal
- Clone:UCHT2
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:Ly-1|Leu-1|T1
- Cat. No.:75843-040
- Supplier no.:09111-80-25
Specifications
About this item
The UCHT2 monoclonal antibody specifically reacts with human CD5, a 67 kda type 1 transmembrane glycoprotein. CD5 is expressed on mature T cells, a subset of B cells, and peripheral blood dendritic cells. B cells that are CD5+ produce mostly IgM polyreactive antibodies. Its ligand is CD72, which is involved in T, B cell proliferation and interaction. The UCT2 antibody also recognizes non-human primate CD5.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human