- Antibody Type:Primary
- Antigen Symbol:CD45.1
- Clonality:Monoclonal
- Clone:A20
- Conjugation:BG Violet 450
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Antigen Synonyms:SJL|T200|LCA|Ly5.1
- Cat. No.:75841-988
- Supplier no.:07522-40-100
The A20 monoclonal antibody reacts with the leukocytes of the CD45.1-expressing mouse strains (DA, SJL/J, RIII, and STS/A). It does not cross-react with cells that express CD45.2. The CD45 molecule is a member of the Protein Tyrosine Phosphatase (PTP) family, because its intracellular region contains two PTP domains. The extracellular region’s variability is caused by different levels of glycosylation, and the splicing of the 4, 5, and 6 exons. The isoforms found in the mouse strains depend on the activation state, maturation stage and cell type, and are very important in B and T lymphocytes antigen receptor signal transduction. The A20 antibody inhibits some of the B lymphocytes responses, from CD45.1-expressing mice, to lipopolysaccharides and antigens. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse