Anti-CD41a Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HIP8]
Catalog # 75842-862
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD41a
- Clonality:Monoclonal
- Clone:HIP8
- Conjugation:PE (Phycoerythrin)
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:Integrin alpha Iib|platelet GPIIb
- Cat. No.:75842-862
- Supplier no.:03511-60-25
Specifications
About this item
The HIP8 monoclonal antibody specifically reacts with human CD41 (GPIIb), a subunit of the calcium-dependent complex with CD61 (IIIa). The complex is expressed on megakaryocytes and platelets. The HIP8 antibody blocks the aggregation of platelets and inhibits platelet activation.
Type: Primary
Antigen: CD41a
Clonality: Monoclonal
Clone: HIP8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human