Anti-CD41 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MWReg30]
Catalog # 75842-866
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Symbol:CD41
- Clonality:Monoclonal
- Clone:MWReg30
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Cat. No.:75842-866
- Supplier no.:03512-50-100
Specifications
About this item
The MWReg30 monoclonal antibody specifically reacts with mouse CD41, a transmembrane glycoprotein also known as integrin alpha 2b GPIIb. It is expressed on hematopoietic progenitors, megakaryocytes, and platelets. It forms a receptor with CD61 (integrin beta 3) that binds adhesion molecules such as fibrinogen, fibronectin, von Willebrand factor, and thrombin. Defects or absence of CD41 has been found to lead to coagulation disorders. The expression profile of CD150+,CD48-, and CD41- can be used to identify hematopoietic stem cells.
Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: MWReg30
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse