Anti-PECAM1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 390]
Catalog # 75842-854
Supplier: BIOGEMS INTERNATIONAL INC.
Specifications
- Antibody Type:Primary
- Antigen Name:Platelet-Endothelial Cell Adhesion Molecule 1
- Antigen Symbol:PECAM1
- Clonality:Monoclonal
- Clone:390
- Conjugation:APC (Allophycocyanin)
- Flow Cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:100 μg
- Form:liquid
- Antigen Synonyms:endoCAM|PECAM-1|platelet and endothelial cell adhesion molecule 1|CD31|GPIIA|CD31/EndoCAM|PECA1
- Cat. No.:75842-854
- Supplier no.:03412-80-100
Specifications
About this item
The 390 monoclonal antibody specifically reacts with mouse cd31, a 130-140 kDA type I transmembrane glycoprotein also known as platelet-endothelial cell adhesion molecule-1 (PECAM-1). CD31 is reported to bind to CD38 and is expressed on platelets, monocytes, granulocytes, and endothelial cells. It plays a role in angiogenesis, wound healing, cellular migration, and signal transduction.
Type: Primary
Antigen: PECAM1
Clonality: Monoclonal
Clone: 390
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse