Specifications
- Antibody Type:Primary
- Antigen Symbol:CD3
- Clonality:Monoclonal
- Clone:OKT3
- Conjugation:BG Violet 450
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:Leu-4|CD3µ|T3
- Cat. No.:75842-902
- Supplier no.:05121-40-25
Specifications
About this item
The OKT3 monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex contains γ, δ, and ε chains, and it is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The OKT3 antibody is an immunosuppressive, which has proven to be an effective therapeutic agent in liver, heart, and renal allograft rejection.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human