Anti-CD2 Mouse Monoclonal Antibody (BG Violet 450) [clone: RPA-2.10]
75841-620
- Antibody Type:Primary
- Antigen Name:Cluster of differentiation 2
- Antigen Symbol:CD2
- Clonality:Monoclonal
- Clone:RPA-2.10
- Conjugation:BG Violet 450
- Flow Cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 tests
- Form:liquid
- Antigen Synonyms:Ly-37|T11|LFA-2|SRBC-R
- Cat. No.:75841-620
- Supplier no.:04111-40-25
The RPA-2.10 monoclonal antibody specifically binds to human CD2, a 50 kDA type I transmembrane glycoprotein. CD2 is expressed on NK cells, thymocytes, T lymphocytes, and B cell subsets. CD2 is involved in cell-cell adhesion, T cell activation, and T cell signaling. Its ligand is CD58 and it is reported to bind to CD48, CD59, and CD15. The RPA-2.10 antibody is reported to be cross-reactive with pigs and non-human primates.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human