Order Entry
Puerto Rico
ContactUsLinkComponent
Bacteria Recombinant Streptavidin (from E. coli)
Bacteria Recombinant Streptavidin (from E. coli)
Supplier:  Anaspec
Certificates
Bacteria Recombinant Streptavidin (from E. coli)
Supplier:  Anaspec

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Bacteria
  • Storage Conditions:
    4 °C
  • Gene ID:
    P02701
  • Protein Synonyms:
    Avidin|Recombinant avidin
  • Protein/Peptide Name:
    Streptavidin
  • Molecular Weight:
    56 kDa

Specifications

Related Information

Frequently Bought Together