About this item
Matrix metalloproteinases (MMP’s) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-1 (collagenase-1) is involved in tumor development and metastasis and rheumatoid arthritis. It is proposed as a therapeutic target for these diseases. MMP-1 digests a broad range of substrates, including α-1 antitrypsin, myelin basic protein, collagen I, II, III, VII, VIII, casein, gelatin, and others
The sequence (Accession # NP_002412) corresponding to the catalytic domain (aa 106-261) of Human MMP-1 was expressed in E. coli. The recombinant human MMP-1 was purified from bacterial lysate and refolded using proprietary technique. The molecular weight of the recombinant Human MMP-1 Catalytic Domain is ~17.5 kDa.
17.5 kDa.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–78 °C
- Protein Synonyms:Matrix metalloproteinase 1|MMP-1|MMP-1 Catalytic Domain
- UniProtKB:P03956
- Protein/Peptide Name:MMP-1
- Purity:>95% as estimated by SDS-PAGE
- Molecular Weight:17.5 kDa