Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant MMP-14 (from E. coli)
Human Recombinant MMP-14 (from E. coli)
  103010-390
 :  
Human Recombinant MMP-14 (from E. coli)
  103010-390
 :  AS-72068
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    100 µl
  • Storage Conditions:
    –78 °C
  • Protein Synonyms:
    Matrix metalloproteinase 14|MMP-14|MMP-14 recombinant
  • UniProtKB:
    P50281
  • Protein/Peptide Name:
    MMP-14
  • Molecular Weight:
    31 kDa
  • Concentration:
    10 μg/mL
  • Cat. No.:
    103010-390
  • Supplier no.:
    AS-72068

 

 

Matrix metalloproteinases (MMPs) belong to a family of secreted or membrane-associated zinc endopeptidases capable of digesting extracellular matrix components. MMP-14 (MT1-MMP), membrane-type MMP, plays an important role in tumor invasion. MMP-14 is expressed on the surface of invasive tumor cells, in stromal cells of human colon, breast, and head and neck carcinomas. MMP-14 is secreted as zymogen with a prodomain, a catalytic domain, a hinge region, a hemopexin-like domain, and a transmembrane domain. It can activate pro-MMP-23 and degrade a variety of substrates, including fibrillar collagens I, II, III, fibronectin, vitronectin and laminin-1.

A truncated human MMP-14 with His-tag was expressed in E. coli. The Mr on SDS-PAGE is 31-kDa. Incubation with 1 mM APMA at 37°C for 2 hr will activate MMP-14. Its activity can be measured by FRET peptides